Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLE27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113494.2 | NP_566783.1 | 1 | 502 | 86 | |
GenBank mRNA | BT028939.1 | - | 1 | 201 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |