Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035697.2 | NP_001030774.1 | 3 - 4 | 1463 | 76 | |
NM_001035699.2 | NP_001030776.2 | 3 - 4 | 1360 | 76 | ||
NM_001338833.1 | NP_001327170.1 | 2 - 3 | 1620 | 76 | ||
NM_001338834.1 | NP_001327171.1 | 3 - 4 | 1438 | 76 | ||
NM_113586.4 | NP_189309.2 | 2 - 3 | 1717 | 76 | ||
GenBank mRNA | AY079016.1 | - | 2 - 3 | 1525 | 76 | |
AY195621.1 | - | 2 - 3 | 1228 | 76 | ||
BT020379.1 | - | 2 - 3 | 1228 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |