Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113620.4 | NP_189342.1 | 1 - 2 | 1072 | 67 | |
GenBank mRNA | AY063837.1 | - | 1 - 2 | 572 | 67 | |
AY117212.1 | - | 1 - 2 | 509 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |