Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338860.1 | NP_001327188.1 | 1 - 2 | 647 | 76 | |
NM_113630.3 | NP_566809.1 | 1 - 2 | 647 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |