Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113645.3 | NP_566815.1 | 1 - 2 | 274 | 68 | |
GenBank mRNA | AK175284.1 | - | 1 - 2 | 103 | 68 | |
AY085053.1 | - | 1 - 2 | 115 | 68 | ||
AY572781.1 | - | 1 - 2 | 91 | 68 | ||
BT024476.1 | - | 1 - 2 | 91 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |