Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125669.3 | NP_201080.1 | 3 - 4 | 483 | 110 | |
GenBank mRNA | AY062850.1 | - | 3 - 4 | 374 | 110 | |
AY086631.1 | - | 3 - 4 | 374 | 110 | ||
AY114572.1 | - | 2 - 3 | 311 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |