Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345564.1 | NP_001331034.1 | 6 - 7 | 966 | 118 | |
NM_125682.5 | NP_201093.1 | 7 - 8 | 877 | 118 | ||
GenBank mRNA | AF079485.1 | - | 7 - 8 | 633 | 118 | |
AK176893.1 | - | 7 - 8 | 684 | 118 | ||
BT002997.1 | - | 7 - 8 | 679 | 118 | ||
BT004459.1 | - | 7 - 8 | 604 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |