Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125754.3 | NP_201164.1 | 1 - 2 | 589 | 91 | |
GenBank mRNA | AK228300.1 | - | 1 - 2 | 420 | 91 | |
BT003134.1 | - | 1 - 2 | 377 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |