Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125754.3 | NP_201164.1 | 2 - 3 | 919 | 79 | |
GenBank mRNA | AK228300.1 | - | 2 - 3 | 750 | 79 | |
BT003134.1 | - | 2 - 3 | 707 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |