Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARI14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125768.1 | NP_201178.1 | 1 - 2 | 839 | 84 | |
GenBank mRNA | DQ056737.1 | - | 1 - 2 | 839 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |