Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGAL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125776.2 | NP_201186.1 | 15 - 16 | 1947 | 78 | |
GenBank mRNA | AY093118.1 | - | 15 - 16 | 1947 | 78 | |
BT010338.1 | - | 15 - 16 | 1779 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |