Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSW3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345613.1 | NP_001330435.1 | 3 - 4 | 2125 | 75 | |
NM_125779.3 | NP_201189.1 | 3 - 4 | 2051 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |