Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UVR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125781.4 | NP_201191.1 | 3 - 4 | 766 | 69 | |
GenBank mRNA | AY125497.1 | - | 368 | 69 | ||
BT000600.1 | - | 2 - 3 | 285 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |