Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345617.1 | NP_001332437.1 | 2 - 3 | 392 | 79 | |
NM_125796.5 | NP_201205.1 | 3 - 4 | 261 | 79 | ||
GenBank mRNA | AY070383.1 | - | 3 - 4 | 248 | 79 | |
AY096575.1 | - | 3 - 4 | 222 | 79 | ||
Z83312.1 | - | 258 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |