Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125800.5 | NP_201209.1 | 2 - 3 | 723 | 79 | |
GenBank mRNA | AY062835.1 | - | 2 - 3 | 480 | 79 | |
AY081594.1 | - | 2 - 3 | 424 | 79 | ||
U32176.1 | - | 2 - 3 | 454 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |