Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125801.4 | NP_201210.1 | 4 - 5 | 779 | 78 | |
GenBank mRNA | AF241841.1 | - | 4 - 5 | 734 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |