Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125831.3 | NP_201240.1 | 2 - 3 | 272 | 84 | |
GenBank mRNA | AY059928.1 | - | 2 - 3 | 175 | 84 | |
AY081564.1 | - | 2 - 3 | 117 | 84 | ||
U96924.1 | - | 2 - 3 | 152 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |