Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125840.4 | NP_201249.2 | 4 - 5 | 452 | 68 | |
GenBank mRNA | AY308736.1 | - | 4 - 5 | 368 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |