Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125849.4 | NP_201258.2 | 2 - 3 | 828 | 106 | |
GenBank mRNA | AK118783.1 | - | 2 - 3 | 523 | 106 | |
BT025746.1 | - | 2 - 3 | 425 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |