Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345677.1 | NP_001331037.1 | 1 - 2 | 346 | 70 | |
NM_125926.4 | NP_201331.1 | 3 - 4 | 700 | 70 | ||
GenBank mRNA | AY058076.1 | - | 3 - 4 | 539 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |