Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCD4;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203691.1 | NP_001190620.1 | 5 - 6 | 922 | 106 | |
NM_001345680.1 | NP_001331197.1 | 4 - 5 | 919 | 106 | ||
NM_125940.3 | NP_201345.1 | 4 - 5 | 919 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |