Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCD4;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203691.1 | NP_001190620.1 | 6 - 7 | 1047 | 97 | |
NM_125940.3 | NP_201345.1 | 5 - 6 | 1044 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |