Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTE7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125961.4 | NP_201366.3 | 2 - 3 | 3003 | 93 | |
GenBank mRNA | AK226877.1 | - | 2 - 3 | 1879 | 93 | |
BT008626.1 | - | 2 - 3 | 1877 | 93 | ||
BT030368.1 | - | 2 - 3 | 1686 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |