Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085334.1 | NP_001078803.1 | 5 - 6 | 991 | 62 | |
NM_001345724.1 | NP_001330475.1 | 6 - 7 | 1155 | 62 | ||
NM_001345725.1 | NP_001330476.1 | 3 - 4 | 795 | 62 | ||
NM_001345726.1 | NP_001330474.1 | 2 - 3 | 624 | 62 | ||
NM_125986.3 | NP_201390.1 | 6 - 7 | 1016 | 62 | ||
GenBank mRNA | AF360256.1 | - | 6 - 7 | 981 | 62 | |
AF528058.1 | - | 5 - 6 | 828 | 62 | ||
AY040070.1 | - | 5 - 6 | 777 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |