Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333259.1 | NP_001322815.1 | 1 - 2 | 1401 | 102 | |
NM_001333260.1 | NP_001322811.1 | 2 - 3 | 1258 | 102 | ||
NM_179446.5 | NP_849777.1 | 2 - 3 | 1263 | 102 | ||
GenBank mRNA | AB017160.1 | - | 2 - 3 | 1178 | 102 | |
AF093545.1 | - | 2 - 3 | 1180 | 102 | ||
BT026443.1 | - | 1 - 2 | 1075 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |