Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198228.1 | NP_001185157.1 | 5 - 6 | 1380 | 63 | |
NM_001333256.1 | NP_001319166.1 | 4 - 5 | 1771 | 63 | ||
NM_001333257.1 | NP_001322813.1 | 3 - 4 | 1923 | 63 | ||
NM_001333258.1 | NP_001322814.1 | 4 - 5 | 1651 | 63 | ||
NM_001333259.1 | NP_001322815.1 | 3 - 4 | 1502 | 63 | ||
NM_001333260.1 | NP_001322811.1 | 4 - 5 | 1359 | 63 | ||
NM_179446.5 | NP_849777.1 | 4 - 5 | 1364 | 63 | ||
GenBank mRNA | AB017160.1 | - | 4 - 5 | 1279 | 63 | |
AF093545.1 | - | 4 - 5 | 1281 | 63 | ||
BT026443.1 | - | 3 - 4 | 1176 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |