Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198036.2 | NP_001184965.1 | 5 - 6 | 2031 | 74 | |
NM_101044.3 | NP_172637.2 | 4 - 5 | 1920 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |