Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198036.2 | NP_001184965.1 | 10 - 11 | 2701 | 98 | |
NM_101044.3 | NP_172637.2 | 9 - 10 | 2590 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |