Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101076.2 | NP_172668.1 | 1 | 2357 | 64 | |
GenBank mRNA | AY026364.1 | - | 1 | 2225 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |