Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KEU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101108.4 | NP_563905.1 | 3 - 4 | 519 | 83 | |
GenBank mRNA | AK227584.1 | - | 3 - 4 | 507 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |