Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035952.3 | NP_001031029.1 | 3 - 4 | 568 | 80 | |
NM_001035953.2 | NP_001031030.1 | 3 - 4 | 575 | 80 | ||
NM_101123.4 | NP_563910.2 | 4 - 5 | 546 | 80 | ||
GenBank mRNA | AK175663.1 | - | 3 - 4 | 562 | 80 | |
AK176660.1 | - | 3 - 4 | 549 | 80 | ||
AK221656.1 | - | 4 - 5 | 466 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |