Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP87A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198045.1 | NP_001184974.1 | 4 - 5 | 1117 | 119 | |
NM_001332045.1 | NP_001320967.1 | 4 - 5 | 1099 | 119 | ||
NM_001332046.1 | NP_001320968.1 | 4 - 5 | 1099 | 119 | ||
NM_101144.5 | NP_172734.2 | 4 - 5 | 1172 | 119 | ||
GenBank mRNA | AK176324.1 | - | 4 - 5 | 1097 | 119 | |
BT010888.1 | - | 4 - 5 | 919 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |