Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DET3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101154.4 | NP_563916.1 | 3 - 4 | 1684 | 81 | |
GenBank mRNA | AB493453.1 | - | 3 - 4 | 305 | 81 | |
AF208261.1 | - | 3 - 4 | 314 | 81 | ||
AF334725.1 | - | 3 - 4 | 305 | 81 | ||
AY059872.1 | - | 3 - 4 | 372 | 81 | ||
AY093334.1 | - | 3 - 4 | 305 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |