Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101159.2 | NP_563919.1 | 4 - 5 | 789 | 73 | |
GenBank mRNA | AY056178.1 | - | 4 - 5 | 716 | 73 | |
AY091264.1 | - | 4 - 5 | 431 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |