Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPA-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123804.1 | NP_001117276.1 | 3 - 4 | 498 | 110 | |
NM_001198048.1 | NP_001184977.1 | 3 - 4 | 614 | 110 | ||
NM_001332055.1 | NP_001321059.1 | 3 - 4 | 628 | 110 | ||
NM_101161.3 | NP_172750.1 | 3 - 4 | 574 | 110 | ||
GenBank mRNA | BT008637.1 | - | 3 - 4 | 485 | 110 | |
BT015867.1 | - | 3 - 4 | 457 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |