Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORP1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198051.1 | NP_001184980.1 | 6 - 7 | 2286 | 68 | |
NM_101187.3 | NP_172776.1 | 6 - 7 | 2303 | 68 | ||
GenBank mRNA | AF462814.1 | - | 6 - 7 | 2219 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |