Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332064.1 | NP_001322783.1 | 6 - 7 | 3247 | 71 | |
NM_101194.2 | NP_563924.1 | 6 - 7 | 3012 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |