Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2AA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035958.1 | NP_001031035.1 | 5 - 6 | 921 | 81 | |
NM_001198052.1 | NP_001184981.1 | 5 - 6 | 974 | 81 | ||
NM_001332065.1 | NP_001320387.1 | 5 - 6 | 1099 | 81 | ||
NM_101203.5 | NP_172790.2 | 5 - 6 | 1020 | 81 | ||
GenBank mRNA | AY099760.1 | - | 5 - 6 | 920 | 81 | |
BT002601.1 | - | 4 - 5 | 831 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |