Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084061.1 | NP_001077530.1 | 8 - 9 | 729 | 74 | |
NM_101214.4 | NP_172801.1 | 8 - 9 | 1044 | 74 | ||
GenBank mRNA | AF410271.1 | - | 8 - 9 | 731 | 74 | |
AY049259.1 | - | 8 - 9 | 731 | 74 | ||
AY090275.1 | - | 7 - 8 | 660 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |