Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084065.3 | NP_001077534.1 | 9 - 10 | 1344 | 73 | |
NM_001332112.1 | NP_001321792.1 | 8 - 9 | 1312 | 73 | ||
NM_001332113.1 | NP_001321793.1 | 7 - 8 | 1101 | 73 | ||
NM_101302.3 | NP_563948.1 | 8 - 9 | 1109 | 73 | ||
GenBank mRNA | AF371982.2 | - | 8 - 9 | 1159 | 73 | |
BT026126.1 | - | 8 - 9 | 931 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |