Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101303.4 | NP_563949.1 | 1 - 2 | 244 | 76 | |
GenBank mRNA | AF332450.1 | - | 1 - 2 | 106 | 76 | |
AY059942.1 | - | 1 - 2 | 229 | 76 | ||
AY114599.1 | - | 1 - 2 | 106 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |