Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TWN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101328.4 | NP_172913.1 | 14 - 15 | 1692 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |