Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNATM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160868.2 | NP_001154340.1 | 1 - 2 | 219 | 71 | |
NM_101345.2 | NP_172929.1 | 1 - 2 | 219 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |