Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DL1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101352.3 | NP_172936.1 | 11 - 12 | 1344 | 80 | |
GenBank mRNA | AJ304841.1 | - | 11 - 12 | 1268 | 80 | |
AY039955.1 | - | 11 - 12 | 1229 | 80 | ||
AY150494.1 | - | 11 - 12 | 1068 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |