Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQD31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106127.5 | NP_177607.2 | 2 - 3 | 400 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |