Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106129.4 | NP_565090.1 | 9 - 10 | 1472 | 75 | |
NM_202414.1 | NP_974143.1 | 9 - 10 | 1471 | 75 | ||
GenBank mRNA | AF367342.1 | - | 9 - 10 | 1470 | 75 | |
AY056055.1 | - | 9 - 10 | 1472 | 75 | ||
AY124864.1 | - | 9 - 10 | 1376 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |