Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334667.1 | NP_001319383.1 | 9 - 10 | 1273 | 71 | |
NM_001334668.1 | NP_001322361.1 | 6 - 7 | 1947 | 71 | ||
NM_001334669.1 | NP_001322362.1 | 7 - 8 | 1863 | 71 | ||
NM_001334670.1 | NP_001322363.1 | 8 - 9 | 1715 | 71 | ||
NM_001334671.1 | NP_001322364.1 | 8 - 9 | 1404 | 71 | ||
NM_001334672.1 | NP_001322360.1 | 9 - 10 | 1285 | 71 | ||
NM_106139.3 | NP_177619.2 | 8 - 9 | 1791 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |