Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH10A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198470.1 | NP_001185399.1 | 8 - 9 | 993 | 76 | |
NM_106150.4 | NP_565094.1 | 8 - 9 | 1101 | 76 | ||
GenBank mRNA | AY087395.1 | - | 8 - 9 | 1008 | 76 | |
AY093071.1 | - | 8 - 9 | 966 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |