Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLP-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106160.3 | NP_177640.1 | 1 - 2 | 285 | 81 | |
GenBank mRNA | AK175343.1 | - | 1 - 2 | 94 | 81 | |
BT030053.1 | - | 1 - 2 | 54 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |