Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334689.1 | NP_001320421.1 | 2 - 3 | 926 | 87 | |
NM_106197.3 | NP_177676.2 | 3 - 4 | 828 | 87 | ||
GenBank mRNA | AB493535.1 | - | 3 - 4 | 735 | 87 | |
DQ446432.1 | - | 3 - 4 | 735 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |